Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUGUUGCUCGUUGAACCCCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Dog | cfa-miR-409 | MIMAT0006726 |
Stem-loop Details
Dog

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | cfa-miR-409 |
---|---|
miRBase Accession Number: | MI0008147 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AAUGUUGCUCGUUGAACCCCU |
Species: | Dog |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUGUUGCUCGUUGAACCCCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Dog | cfa-miR-409 | MIMAT0006726 |