Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUGUUGCUCGUUGAACCCCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dog | cfa-miR-409 | MIMAT0006726 |
Stem-loop Details
Dog
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cfa-miR-409 |
|---|---|
| miRBase Accession Number: | MI0008147 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAUGUUGCUCGUUGAACCCCU |
| Species: | Dog |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUGUUGCUCGUUGAACCCCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dog | cfa-miR-409 | MIMAT0006726 |