Assay Details
Gene Family ID: | MIPF0000074, mir-105 |
Mature miRNA Sequence: | UCAAAUGCUCAGACUCCUUG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Dog | cfa-miR-105b | MIMAT0010198 |
Pig | ssc-miR-105-2 | MIMAT0002117 |
Stem-loop Details
Dog
Pig
