Assay Details
Gene Family ID: | MIPF0000074, mir-105 |
Mature miRNA Sequence: | UCAAAUGCUCAGACUCCUUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dog | cfa-miR-105b | MIMAT0010198 |
| Pig | ssc-miR-105-2 | MIMAT0002117 |
Stem-loop Details
Dog
Pig
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cfa-miR-105b |
|---|---|
| miRBase Accession Number: | MI0010378 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCAAAUGCUCAGACUCCUUG |
| Species: | Dog, Pig |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000074, mir-105 |
Mature miRNA Sequence: | UCAAAUGCUCAGACUCCUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dog | cfa-miR-105b | MIMAT0010198 |
| Pig | ssc-miR-105-2 | MIMAT0002117 |