Assay Details
Mature miRNA Sequence: | AGAUGCAAGAGAAUUUUCAGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-1339-5p | MIMAT0006861 |
Stem-loop Details
Ornithorhynchus anatinus
More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-1339 (v18)

Genomic Map