Assay Details
Mature miRNA Sequence: | AGAUGCAAGAGAAUUUUCAGU | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-1339-5p | MIMAT0006861 | 
Stem-loop Details
Ornithorhynchus anatinus
More Information
Related Products
miRBase Alias:
						
							Ochrobactrum anthropi: oan-miR-1339 (v18)
						
					
				
Genomic Map