Assay Details
| Gene Family ID: | MIPF0000179, mir-458 | 
| Mature miRNA Sequence: | AGCGCCAUUUUCAGAGCUAU | 
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-458-5p | MIMAT0007009 | 
Stem-loop Details
Ornithorhynchus anatinus
 Back To Top
 Back To TopMore Information
Related Products
miRBase Alias:
						
							Ochrobactrum anthropi: oan-miR-458* (v18)
						
					
				 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top mirVana® miRNA inhibitor :
						mirVana® miRNA inhibitor :