Assay Details
Gene Family ID: | MIPF0000179, mir-458 |
Mature miRNA Sequence: | AGCGCCAUUUUCAGAGCUAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-458-5p | MIMAT0007009 |
Stem-loop Details
Ornithorhynchus anatinus
More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-458* (v18)

Genomic Map