Assay Details
Mature miRNA Sequence: | GUAAGUGACCCAGUUGAGCG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila simulans | dsi-miR-2579 | MIMAT0012512 |
Stem-loop Details
Drosophila simulans

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | dsi-miR-2579 |
---|---|
miRBase Accession Number: | MI0011781 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GUAAGUGACCCAGUUGAGCG |
Species: | Drosophila simulans |
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | GUAAGUGACCCAGUUGAGCG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila simulans | dsi-miR-2579 | MIMAT0012512 |