Assay Details
Mature miRNA Sequence: | GUAAGUGACCCAGUUGAGCG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila simulans | dsi-miR-2579 | MIMAT0012512 |
Stem-loop Details
Drosophila simulans
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | dsi-miR-2579 |
|---|---|
| miRBase Accession Number: | MI0011781 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUAAGUGACCCAGUUGAGCG |
| Species: | Drosophila simulans |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | GUAAGUGACCCAGUUGAGCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila simulans | dsi-miR-2579 | MIMAT0012512 |