Assay Details
Gene Family ID: | MIPF0001912, mir-2217 |
Mature miRNA Sequence: | UCGACCCUUGUGCCUGUUUCGGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Caenorhabditis elegans | cel-miR-2217a-3p | MIMAT0011452 |
Stem-loop Details
Caenorhabditis elegans

More Information
Related Products
miRBase Alias:
Caenorhabditis elegans: cel-miR-2217-3p (v20)
Caenorhabditis elegans: cel-miR-2217 (v17)
Caenorhabditis elegans: cel-miR-2217 (v17)
