Assay Details
Gene Family ID: | MIPF0001912, mir-2217 |
Mature miRNA Sequence: | UCGACCCUUGUGCCUGUUUCGGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-2217a-3p | MIMAT0011452 |
Stem-loop Details
Caenorhabditis elegans
More Information
Related Products
miRBase Alias:
Caenorhabditis elegans: cel-miR-2217-3p (v20)
Caenorhabditis elegans: cel-miR-2217 (v17)
Caenorhabditis elegans: cel-miR-2217 (v17)

Genomic Map