Assay Details
Gene Family ID: | MIPF0000436, mir-544 |
Mature miRNA Sequence: | AUUCUGCAUUUUUAACAAGUUC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Horse | eca-miR-544b | MIMAT0013118 |
Stem-loop Details
Horse

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | eca-miR-544b |
---|---|
miRBase Accession Number: | MI0012868 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AUUCUGCAUUUUUAACAAGUUC |
Species: | Horse |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000436, mir-544 |
Mature miRNA Sequence: | AUUCUGCAUUUUUAACAAGUUC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Horse | eca-miR-544b | MIMAT0013118 |