Assay Details
| Gene Family ID: | MIPF0000436, mir-544 | 
| Mature miRNA Sequence: | AUUCUGCAUUUUUAACAAGUUC | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Horse | eca-miR-544b | MIMAT0013118 | 
Stem-loop Details
Horse
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | eca-miR-544b | 
|---|---|
| miRBase Accession Number: | MI0012868 | 
| miRBase Version: | v22.1 | 
| Mature miRNA Sequence: | AUUCUGCAUUUUUAACAAGUUC | 
| Species: | Horse | 
| Product Type: | mirVana® miRNA inhibitor | 
| Gene Family ID: | MIPF0000436, mir-544 | 
| Mature miRNA Sequence: | AUUCUGCAUUUUUAACAAGUUC | 
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Horse | eca-miR-544b | MIMAT0013118 | 
 Back To Top
 Back To Top