Assay Details
Gene Family ID: | MIPF0000985, mir-1013 |
Mature miRNA Sequence: | AUAAAAGUAUGCCGAACUCGCAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila simulans | dsi-miR-1013 | MIMAT0012499 |
Stem-loop Details
Drosophila simulans
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | dsi-miR-1013 |
|---|---|
| miRBase Accession Number: | MI0011770 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUAAAAGUAUGCCGAACUCGCAG |
| Species: | Drosophila simulans |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000985, mir-1013 |
Mature miRNA Sequence: | AUAAAAGUAUGCCGAACUCGCAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila simulans | dsi-miR-1013 | MIMAT0012499 |