Assay Details
Gene Family ID: | MIPF0000985, mir-1013 |
Mature miRNA Sequence: | AUAAAAGUAUGCCGAACUCGCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila simulans | dsi-miR-1013 | MIMAT0012499 |
Stem-loop Details
Drosophila simulans
