Assay Details
Gene Family ID: | MIPF0000715, mir-1175 |
Mature miRNA Sequence: | AAGUGGAGCAGUGGUCUCUUCAC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Tribolium castaneum | tca-miR-1175-5p | MIMAT0018818 |
Stem-loop Details
Tribolium castaneum

Genomic Map