Assay Details
Gene Family ID: | MIPF0000715, mir-1175 |
Mature miRNA Sequence: | AAGUGGAGCAGUGGUCUCUUCAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Tribolium castaneum | tca-miR-1175-5p | MIMAT0018818 |
Stem-loop Details
Tribolium castaneum
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | tca-miR-1175-5p |
|---|---|
| miRBase Accession Number: | MI0016357 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAGUGGAGCAGUGGUCUCUUCAC |
| Species: | Tribolium castaneum |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000715, mir-1175 |
Mature miRNA Sequence: | AAGUGGAGCAGUGGUCUCUUCAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Tribolium castaneum | tca-miR-1175-5p | MIMAT0018818 |