Assay Details
Gene Family ID: | MIPF0000082, mir-193 |
Mature miRNA Sequence: | AGGGUCUUGGCGGUCUAGUGGG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Heliconius melpomene | hme-miR-193-5p | MIMAT0020538 |
Stem-loop Details
Heliconius melpomene

More Information
Related Products
miRBase Alias:
Human metapneumovirus: hme-miR-193* (v18)
