Assay Details
Gene Family ID: | MIPF0000082, mir-193 |
Mature miRNA Sequence: | AGGGUCUUGGCGGUCUAGUGGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Heliconius melpomene | hme-miR-193-5p | MIMAT0020538 |
Stem-loop Details
Heliconius melpomene
More Information
Related Products
miRBase Alias:
Human metapneumovirus: hme-miR-193* (v18)

Genomic Map