Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAGGUAAUUGCAGUUUUUCCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548ai | MIMAT0018989 |
| Human | hsa-miR-570-5p | MIMAT0022707 |
Stem-loop Details
Human

Genomic Map