Assay Details
Amplicon Length: | 112 |
Chromosome Location: | Chr. 3 - 195699705 - 195699705 [+] on 195699705 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-570-5p | MIMAT0022707 |
| Human | hsa-miR-570-3p | MIMAT0003235 |
Stem-loop Details
Human

Genomic Map