Assay Details
| Gene Family ID: | MIPF0000293, mir-67 | 
| Mature miRNA Sequence: | CCACAACCUCCUUGAGUGAGCGA | 
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Acyrthosiphon pisum | api-miR-307 | MIMAT0014729 | 
Stem-loop Details
Acyrthosiphon pisum
 Back To Top
 Back To TopMore Information
Updated Mapping Information
 This assay detects a miRNA target from an earlier version of miRbase that has been obsoleted in miRbase v.22. This assay is still available for purchase for continuity of studies. For full target details including sequence, please refer to the miRbase database at http://www.mirbase.org/.
					            	This assay detects a miRNA target from an earlier version of miRbase that has been obsoleted in miRbase v.22. This assay is still available for purchase for continuity of studies. For full target details including sequence, please refer to the miRbase database at http://www.mirbase.org/.Related Products
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top mirVana® miRNA inhibitor :
						mirVana® miRNA inhibitor :