Assay Details
Gene Family ID: | MIPF0000293, mir-67 |
Mature miRNA Sequence: | CCACAACCUCCUUGAGUGAGCGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Acyrthosiphon pisum | api-miR-307 | MIMAT0014729 |
Stem-loop Details
Acyrthosiphon pisum

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | api-miR-307 |
---|---|
miRBase Accession Number: | MI0013930 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CCACAACCUCCUUGAGUGAGCGA |
Species: | Acyrthosiphon pisum |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000293, mir-67 |
Mature miRNA Sequence: | CCACAACCUCCUUGAGUGAGCGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Acyrthosiphon pisum | api-miR-307 | MIMAT0014729 |