Assay Details
Gene Family ID: | MIPF0000139, mir-500 | 
Mature miRNA Sequence: | GCACCCAGGCAAGGAUUCUG | 
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-500b-3p | MIMAT0027032 | 
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | hsa-miR-500b-3p | 
|---|---|
| miRBase Accession Number: | MI0015903 | 
| miRBase Version: | v22.1 | 
| Mature miRNA Sequence: | GCACCCAGGCAAGGAUUCUG | 
| Species: | Human | 
| Product Type: | TaqMan™ MicroRNA Assay | 
Gene Family ID: | MIPF0000139, mir-500 | 
Mature miRNA Sequence: | GCACCCAGGCAAGGAUUCUG | 
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-500b-3p | MIMAT0027032 |