Assay Details
Amplicon Length: | 85 |
Chromosome Location: | Chr. X - 50010918 - 50010918 [-] on 50010918 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-500b-5p | MIMAT0016925 |
Human | hsa-miR-500b-3p | MIMAT0027032 |
Stem-loop Details
Human
