Assay Details
Gene Family ID: | MIPF0000112, mir-134 |
Mature miRNA Sequence: | CCUGUGGGCCACCUAGUCACCAA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-134-3p | MIMAT0026481 |
| Rhesus monkey | mml-miR-134-3p | MIMAT0026819 |
Stem-loop Details
Human
Rhesus monkey

Genomic Map