Assay Details
Amplicon Length: | 58 |
Chromosome Location: | Chr. 14 - 101054506 - 101054506 [+] on 101054506 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-134-5p | MIMAT0000447 |
| Human | hsa-miR-134-3p | MIMAT0026481 |
| Human | hsa-miR-382-5p | MIMAT0000737 |
| Human | hsa-miR-382-3p | MIMAT0022697 |
Stem-loop Details
Human

Genomic Map