Assay Details
Gene Family ID: | MIPF0000401, mir-874 |
Mature miRNA Sequence: | CGGCCCCACGCACCAGGGUAAGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-874-5p | MIMAT0026718 |
| Dasypus novemcinctus | dno-miR-874-5p | MIMAT0048018 |
| Goat | chi-miR-874-5p | MIMAT0036296 |
| Guinea pig | cpo-miR-874-5p | MIMAT0047393 |
| Rabbit | ocu-miR-874-5p | MIMAT0048575 |
| Rhesus monkey | mml-miR-874-5p | MIMAT0026910 |
Stem-loop Details
Human
Dasypus novemcinctus
Goat
Guinea pig
Rabbit
Rhesus monkey

Genomic Map