Assay Details
Amplicon Length: | 86 |
Chromosome Location: | Chr. 5 - 137647377 - 137647377 [-] on 137647377 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-874-3p | MIMAT0004911 |
| Human | hsa-miR-874-5p | MIMAT0026718 |
Stem-loop Details
Human

Genomic Map