Assay Details
Gene Family ID: | MIPF0001717, mir-1824 |
Mature miRNA Sequence: | UGGCAGUGCCGAUUCUAGUGGC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Caenorhabditis brenneri | cbn-miR-1824a | MIMAT0029736 |
Stem-loop Details
Caenorhabditis brenneri
