Assay Details
Gene Family ID: | MIPF0001717, mir-1824 |
Mature miRNA Sequence: | UGGCAGUGCCGAUUCUAGUGGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis brenneri | cbn-miR-1824a | MIMAT0029736 |
Stem-loop Details
Caenorhabditis brenneri

Genomic Map