Assay Details
Mature miRNA Sequence: | ACCGUGGCUUUAGAUUGUUACU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Astatotilapia burtoni | abu-miR-132-5p | MIMAT0042179 | |
| Chicken | gga-miR-132a-5p | MIMAT0049950 | |
| Chrysemys picta | cpi-miR-132-5p | MIMAT0037748 | |
| Gadus morhua | gmo-miR-132-2-5p | MIMAT0044281 | |
| Ictalurus punctatus | ipu-miR-132a | MIMAT0029405 | |
| Metriaclima zebra | mze-miR-132b | MIMAT0042405 | |
| View More Rows | |||
| Neolamprologus brichardi | nbr-miR-132b | MIMAT0042588 | |
| Ophiophagus hannah | oha-miR-132-5p | MIMAT0036694 | |
| Oreochromis niloticus | oni-miR-132b-5p | MIMAT0042783 | |
| Pundamilia nyererei | pny-miR-132-2 | MIMAT0042979 | |
| Python bivittatus | pbv-miR-132-5p | MIMAT0038933 | |
| Salmo salar | ssa-miR-132-5p | MIMAT0032331 | |
Stem-loop Details
Astatotilapia burtoni
Chicken
Chrysemys picta
Gadus morhua
Ictalurus punctatus
Metriaclima zebra
Neolamprologus brichardi
Ophiophagus hannah
Oreochromis niloticus
Pundamilia nyererei
Python bivittatus
Salmo salar

Genomic Map