Assay Details
Mature miRNA Sequence: | ACCGUGGCUUUAGAUUGUUACU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Astatotilapia burtoni | abu-miR-132-5p | MIMAT0042179 |
Chicken | gga-miR-132a-5p | MIMAT0049950 |
Chrysemys picta | cpi-miR-132-5p | MIMAT0037748 |
Gadus morhua | gmo-miR-132-2-5p | MIMAT0044281 |
Ictalurus punctatus | ipu-miR-132a | MIMAT0029405 |
Metriaclima zebra | mze-miR-132b | MIMAT0042405 |
View More Rows | ||
Neolamprologus brichardi | nbr-miR-132b | MIMAT0042588 |
Ophiophagus hannah | oha-miR-132-5p | MIMAT0036694 |
Oreochromis niloticus | oni-miR-132b-5p | MIMAT0042783 |
Pundamilia nyererei | pny-miR-132-2 | MIMAT0042979 |
Python bivittatus | pbv-miR-132-5p | MIMAT0038933 |
Salmo salar | ssa-miR-132-5p | MIMAT0032331 |
Stem-loop Details
Astatotilapia burtoni
Chicken
Chrysemys picta
Gadus morhua
Ictalurus punctatus
Metriaclima zebra
Neolamprologus brichardi
Ophiophagus hannah
Oreochromis niloticus
Pundamilia nyererei
Python bivittatus
Salmo salar
