Assay Details
Gene Family ID: | MIPF0000044, mir-219 |
Mature miRNA Sequence: | AGUUGGGUCUGGACAUCCCG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Monodelphis domestica | mdo-miR-219-2-3p | MIMAT0031109 |
Stem-loop Details
Monodelphis domestica

Genomic Map