Assay Details
Gene Family ID: | MIPF0000044, mir-219 |
Mature miRNA Sequence: | AGUUGGGUCUGGACAUCCCG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Monodelphis domestica | mdo-miR-219-2-3p | MIMAT0031109 |
Stem-loop Details
Monodelphis domestica
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mdo-miR-219-2-3p |
|---|---|
| miRBase Accession Number: | MI0023783 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGUUGGGUCUGGACAUCCCG |
| Species: | Monodelphis domestica |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000044, mir-219 |
Mature miRNA Sequence: | AGUUGGGUCUGGACAUCCCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Monodelphis domestica | mdo-miR-219-2-3p | MIMAT0031109 |