Assay Details
Gene Family ID: | MIPF0000635, mir-1329 |
Mature miRNA Sequence: | CCCCGUAGCUUGAUCACCAUCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Ophiophagus hannah | oha-miR-1329-3p | MIMAT0036697 |
Python bivittatus | pbv-miR-1329-3p | MIMAT0039076 |
Stem-loop Details
Ophiophagus hannah
Python bivittatus
