Assay Details
Gene Family ID: | MIPF0000635, mir-1329 |
Mature miRNA Sequence: | CCCCGUAGCUUGAUCACCAUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ophiophagus hannah | oha-miR-1329-3p | MIMAT0036697 |
| Python bivittatus | pbv-miR-1329-3p | MIMAT0039076 |
Stem-loop Details
Ophiophagus hannah
Python bivittatus

Genomic Map