Assay Details
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | AACAUCACUUUAAGUCUGUGCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Alligator mississippiensis | ami-miR-499-3p | MIMAT0038302 | |
Anolis carolinensis | aca-miR-499-3p | MIMAT0021981 | |
Chicken | gga-miR-499-3p | MIMAT0026634 | |
Chrysemys picta | cpi-miR-499-3p | MIMAT0037902 | |
Columba livia | cli-miR-499-3p | MIMAT0038661 | |
Ophiophagus hannah | oha-miR-499-3p | MIMAT0036939 | |
View More Rows | |||
Python bivittatus | pbv-miR-499-3p | MIMAT0039066 | |
Salmo salar | ssa-miR-499b-3p | MIMAT0032662 | |
Taeniopygia guttata | tgu-miR-499-3p | MIMAT0027340 | |
Zebrafish | dre-miR-499-3p | MIMAT0032009 |
Stem-loop Details
Alligator mississippiensis
Anolis carolinensis
Chicken
Chrysemys picta
Columba livia
Ophiophagus hannah
Python bivittatus
Salmo salar
Taeniopygia guttata
Zebrafish
