Assay Details
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | AACAUCACUUUAAGUCUGUGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Alligator mississippiensis | ami-miR-499-3p | MIMAT0038302 |
| Anolis carolinensis | aca-miR-499-3p | MIMAT0021981 |
| Chicken | gga-miR-499-3p | MIMAT0026634 |
| Chrysemys picta | cpi-miR-499-3p | MIMAT0037902 |
| Columba livia | cli-miR-499-3p | MIMAT0038661 |
| Ophiophagus hannah | oha-miR-499-3p | MIMAT0036939 |
| View More Rows | ||
| Python bivittatus | pbv-miR-499-3p | MIMAT0039066 |
| Salmo salar | ssa-miR-499b-3p | MIMAT0032662 |
| Taeniopygia guttata | tgu-miR-499-3p | MIMAT0027340 |
| Zebrafish | dre-miR-499-3p | MIMAT0032009 |
Stem-loop Details
Alligator mississippiensis
Anolis carolinensis
Chicken
Chrysemys picta
Columba livia
Ophiophagus hannah
Python bivittatus
Salmo salar
Taeniopygia guttata
Zebrafish

Genomic Map