Assay Details
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | GAAGUUGCCCAUGUUAUUUUCG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-495-5p | MIMAT0022924 |
| Dasypus novemcinctus | dno-miR-495-5p | MIMAT0047930 |
| Pteropus alecto | pal-miR-495-5p | MIMAT0040185 |
| Rabbit | ocu-miR-495-5p | MIMAT0048483 |
| Rhesus monkey | mml-miR-495-5p | MIMAT0026886 |
Stem-loop Details
Human
Dasypus novemcinctus
Pteropus alecto
Rabbit
Rhesus monkey

Genomic Map