Assay Details
Amplicon Length: | 61 |
Chromosome Location: | Chr. 14 - 101033554 - 101033554 [-] on 101033554 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-495-3p | MIMAT0002817 |
| Human | hsa-miR-495-5p | MIMAT0022924 |
Stem-loop Details
Human

Genomic Map