Assay Details
Mature miRNA Sequence: | UGGAAGUGAAUGGUGACUGAG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Cyprinus carpio | ccr-miR-2192 | MIMAT0026328 |
Stem-loop Details
Cyprinus carpio

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ccr-miR-2192 |
---|---|
miRBase Accession Number: | MI0023424 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UGGAAGUGAAUGGUGACUGAG |
Species: | Cyprinus carpio |
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UGGAAGUGAAUGGUGACUGAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Cyprinus carpio | ccr-miR-2192 | MIMAT0026328 |