Assay Details
Mature miRNA Sequence: | UGGAAGUGAAUGGUGACUGAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Cyprinus carpio | ccr-miR-2192 | MIMAT0026328 |
Stem-loop Details
Cyprinus carpio
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ccr-miR-2192 |
|---|---|
| miRBase Accession Number: | MI0023424 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAAGUGAAUGGUGACUGAG |
| Species: | Cyprinus carpio |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UGGAAGUGAAUGGUGACUGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Cyprinus carpio | ccr-miR-2192 | MIMAT0026328 |