Assay Details
Gene Family ID: | MIPF0000483, mir-1271 |
Mature miRNA Sequence: | AGUGCCUGCUAUGUGCCAGGCA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1271-3p | MIMAT0022712 |
| Guinea pig | cpo-miR-1271-3p | MIMAT0047412 |
Stem-loop Details
Human
Guinea pig

Genomic Map