Assay Details
Amplicon Length: | 60 |
Chromosome Location: | Chr. 5 - 176367744 - 176367744 [+] on 176367744 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1271-5p | MIMAT0005796 |
| Human | hsa-miR-1271-3p | MIMAT0022712 |
Stem-loop Details
Human

Genomic Map