Assay Details
Mature miRNA Sequence: | AACAUCACUUUAAGUCUGUGC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Gadus morhua | gmo-miR-499-3p | MIMAT0044197 |
Oryzias latipes | ola-miR-499 | MIMAT0022548 |
Stem-loop Details
Gadus morhua
Oryzias latipes
