Assay Details
Mature miRNA Sequence: | AACAUCACUUUAAGUCUGUGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gadus morhua | gmo-miR-499-3p | MIMAT0044197 |
| Oryzias latipes | ola-miR-499 | MIMAT0022548 |
Stem-loop Details
Gadus morhua
Oryzias latipes
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | gmo-miR-499-3p |
|---|---|
| miRBase Accession Number: | MI0036115 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AACAUCACUUUAAGUCUGUGC |
| Species: | Gadus morhua, Oryzias latipes |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | AACAUCACUUUAAGUCUGUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gadus morhua | gmo-miR-499-3p | MIMAT0044197 |
| Oryzias latipes | ola-miR-499 | MIMAT0022548 |