Assay Details
Mature miRNA Sequence: | UUACAGGCUAUGCUAAUCUAUGC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Salmo salar | ssa-miR-2187-3p | MIMAT0032521 |
Stem-loop Details
Salmo salar

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ssa-miR-2187-3p |
---|---|
miRBase Accession Number: | MI0026616 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UUACAGGCUAUGCUAAUCUAUGC |
Species: | Salmo salar |
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UUACAGGCUAUGCUAAUCUAUGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Salmo salar | ssa-miR-2187-3p | MIMAT0032521 |