Assay Details
Mature miRNA Sequence: | UUACAGGCUAUGCUAAUCUAUGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Salmo salar | ssa-miR-2187-3p | MIMAT0032521 |
Stem-loop Details
Salmo salar
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ssa-miR-2187-3p |
|---|---|
| miRBase Accession Number: | MI0026616 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUACAGGCUAUGCUAAUCUAUGC |
| Species: | Salmo salar |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UUACAGGCUAUGCUAAUCUAUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Salmo salar | ssa-miR-2187-3p | MIMAT0032521 |